missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxocyclopentanecarboxylate, 97+%

Catalog No. AAL0452814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0452814 25 g
AAL0452822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0452814 Supplier Thermo Scientific Chemicals Supplier No. L0452814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-10-9 | C8H12O3 | 156.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-10-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00001412
InChI Key JHZPNBKZPAWCJD-UHFFFAOYSA-N
Synonym ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate
PubChem CID 69136
IUPAC Name ethyl 2-oxocyclopentane-1-carboxylate
SMILES CCOC(=O)C1CCCC1=O

Specifications

Density 1.079
Boiling Point 225°C to 228°C
Flash Point 77°C (170°F)
Refractive Index 1.452
Quantity 25 g
Beilstein 387787
Formula Weight 156.18
Percent Purity ≥97%
Chemical Name or Material Ethyl 2-oxocyclopentanecarboxylate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-253-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only