missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Oxocyclopentaneacetate 95.0+%, TCI America™

Catalog No. p-7129237
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
E09431G 1 g
E09435G 5 g
2 options

Catalog No. E09431G

Supplier: TCI America E09431G

Only null left

Chemical Identifiers

CAS 20826-94-2
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00044715
InChI Key PJMKFKUFBDXYEC-SSDOTTSWSA-N
Synonym ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate
PubChem CID 2723663
IUPAC Name ethyl 2-[(1R)-2-oxocyclopentyl]acetate
SMILES CCOC(=O)C[C@H]1CCCC1=O

Specifications

Color Yellow
Boiling Point 106°C
Quantity 1 g
Formula Weight 170.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Oxocyclopentaneacetate
TSCA No
Recommended Storage Refrigerator