missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Oxocyclopentaneacetate 95.0+%, TCI America™

Catalog No. E09435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09435G 5 g
E09431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09435G Supplier TCI America Supplier No. E09435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20826-94-2
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00044715
InChI Key PJMKFKUFBDXYEC-SSDOTTSWSA-N
Synonym ethyl 2-oxocyclopentylacetate, ethyl 2-2-oxocyclopentyl acetate, ethyl 2-oxocyclopentaneacetate, ethyl 2-oxocyclopentyl acetate, 2-oxocyclopentaneacetic acid ethyl ester, cyclopentaneacetic acid, 2-oxo-, ethyl ester, pubchem23864, ksc201q9p, ethyl 2-oxo-cyclopentyl acetate, ehtyl 2-2-oxocyclopentyl acetate
PubChem CID 2723663
IUPAC Name ethyl 2-[(1R)-2-oxocyclopentyl]acetate
SMILES CCOC(=O)C[C@H]1CCCC1=O

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
Formula Weight 170.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Oxocyclopentaneacetate
TSCA No
Recommended Storage Refrigerator