missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Oxocyclohexanecarboxylate 95.0+%, TCI America™

Catalog No. C091325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C091325ML 25 mL
C0913100ML 100 mL
C0913500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C091325ML Supplier TCI America Supplier No. C091325ML

Chemical Identifiers

CAS 1655-07-8
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00001631
InChI Key FGSGHBPKHFDJOP-UHFFFAOYNA-N
Synonym ethyl 2-oxocyclohexanecarboxylate, ethyl 2-cyclohexanonecarboxylate, 2-carbethoxycyclohexanone, ethyl cyclohexanone-2-carboxylate, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-oxocyclohexanecarboxylic acid ethyl ester, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate
PubChem CID 95543
IUPAC Name ethyl 2-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCCCC1=O

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 170.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Oxocyclohexanecarboxylate
EINECSNumber (9)-0294&(9)-2106
TSCA Yes