missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxocyclohexanecarboxylate, 95%

Catalog No. AAH5949514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5949514 25 g
AAH5949522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5949514 Supplier Thermo Scientific Chemicals Supplier No. H5949514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1655-07-8 | C9H14O3 | 170.21 g/mol

Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 1655-07-8
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00001631
InChI Key FGSGHBPKHFDJOP-UHFFFAOYNA-N
Synonym ethyl 2-oxocyclohexanecarboxylate, ethyl 2-cyclohexanonecarboxylate, 2-carbethoxycyclohexanone, ethyl cyclohexanone-2-carboxylate, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-oxocyclohexanecarboxylic acid ethyl ester, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate
PubChem CID 95543
IUPAC Name ethyl 2-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCCCC1=O

Specifications

Density 1.064
Boiling Point 106°C (11 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.4775
Linear Formula (O=)C6H9CO2C2H5
Quantity 25 g
Beilstein 608356
Solubility Information Soluble in water.
Formula Weight 170.21
Percent Purity 95%
Chemical Name or Material Ethyl 2-oxocyclohexanecarboxylate
Show More Show Less
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 216-731-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only