missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylquinoline-6-carboxylate, 97%

Catalog No. AAH6321303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6321303 1 g
AAH6321306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6321303 Supplier Thermo Scientific Chemicals Supplier No. H6321303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 855763-77-8 | C13H13NO2 | 215.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 855763-77-8
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD09879697
InChI Key WLKKUWXLOSMSMH-UHFFFAOYSA-N
Synonym 2-methylquinoline-6-carboxylic acid ethyl ester, ethyl 2-methyl-6-quinolinecarboxylate, ethyl2-methylquinoline-6-carboxylate, pubchem20820, 2-methylquinoline-6-carboxylic acidethylester, 2-methyl-6-quinolinecarboxylic acid ethyl ester, 6-quinolinecarboxylicacid, 2-methyl-, ethyl ester
PubChem CID 29949719
IUPAC Name ethyl 2-methylquinoline-6-carboxylate
SMILES CCOC(=O)C1=CC2=C(C=C1)N=C(C=C2)C

Specifications

Quantity 1 g
Formula Weight 215.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 2-methylquinoline-6-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only