missing translation for 'onlineSavingsMsg'
Learn More

Éthyle 2-méthylnicotinate 98,0+%, TCI America™

Numéro de catalogue. E08605G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E08605G 5 g
E086025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E08605G Fournisseur TCI America Code fournisseur E08605G

Identifiants chimiques

CAS 1721-26-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00006336
InChI Key TZORNSWZUZDUCU-UHFFFAOYSA-N
Synonym ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727
PubChem CID 74401
IUPAC Name Éthyle 2-méthylpyridine-3-carboxylate
SMILES CCOC(=O)C1=CC=CN=C1C

Spécifications

Color Jaune
Boiling Point 125°C
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquide
Chemical Name or Material Ethyl 2-Methylnicotinate