Learn More
Ethyl 2-methylnicotinate, 97%
CAS: 1721-26-2 | C9H11NO2 | 165.19 g/mol
$121.64 - $381.30
Chemical Identifiers
| CAS | 1721-26-2 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.19 |
| MDL Number | MFCD00006336 |
| InChI Key | TZORNSWZUZDUCU-UHFFFAOYSA-N |
| Synonym | ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727 |
| PubChem CID | 74401 |
| IUPAC Name | ethyl 2-methylpyridine-3-carboxylate |
| SMILES | CCOC(=O)C1=CC=CN=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0325606
|
Thermo Scientific Chemicals
L0325606 |
5 g |
Each for $121.64
|
|
|||||
|
AAL0325614
|
Thermo Scientific Chemicals
L0325614 |
25 g |
Each for $381.30
|
|
|||||
Description
Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 1721-26-2 | |
| 165.19 | |
| TZORNSWZUZDUCU-UHFFFAOYSA-N | |
| 74401 | |
| CCOC(=O)C1=CC=CN=C1C |
| C9H11NO2 | |
| MFCD00006336 | |
| ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727 | |
| ethyl 2-methylpyridine-3-carboxylate |
Specifications
| 1721-26-2 | |
| 107°C to 108°C (13 mmHg) | |
| Recognizable | |
| 1.505 | |
| 5 g | |
| ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727 | |
| TZORNSWZUZDUCU-UHFFFAOYSA-N | |
| ethyl 2-methylpyridine-3-carboxylate | |
| 74401 | |
| 97% |
| 1.07 | |
| 102°C (215°F) | |
| C9H11NO2 | |
| MFCD00006336 | |
| 118200 | |
| Slightly soluble in water. | |
| CCOC(=O)C1=CC=CN=C1C | |
| 165.19 | |
| 165.19 | |
| Ethyl 2-methylnicotinate |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-013-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only