missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylbutyrate, 99%

Catalog No. AC250260050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC250260050 5 g Glass bottle
AC250261000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC250260050 Supplier Thermo Scientific Chemicals Supplier No. 250260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7452-79-1 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7452-79-1
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00012217
InChI Key HCRBXQFHJMCTLF-UHFFFAOYSA-N
Synonym ethyl 2-methylbutyrate, butanoic acid, 2-methyl-, ethyl ester, butyric acid, 2-methyl-, ethyl ester, ethyl 2-methyl butyrate, ethyl alpha-methylbutyrate, dl-2-methylbutyric acid ethyl ester, ethyl 2-methyl-butanoate, ethyl .alpha.-methylbutyrate, 2-methylbutanoic acid ethyl ester, butyric acid, 2-methyl-, ethyl ester 8ci
PubChem CID 24020
IUPAC Name ethyl 2-methylbutanoate
SMILES CCC(C)C(=O)OCC

Specifications

Acidity 1mg KOH/g max.
Color Colorless
Density 0.8750g/mL
Boiling Point 132°C to 133°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3960 to 1.399
Quantity 5 g
Specific Gravity 0.875
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,ether and benzene,soluble in propylene glycol,miscible in fixed oils
Formula Weight 130.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 2-methylbutyrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-225-4
TSCA TSCA

RUO – Research Use Only