missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-Methylbenzoyl)acetate 98.0+%, TCI America™

Catalog No. E11251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11251G Supplier TCI America Supplier No. E11251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51725-82-7
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD03424817
InChI Key UNULPFKXRJPSCO-UHFFFAOYSA-N
Synonym (2-Methylbenzoyl)acetic Acid Ethyl Ester, Ethyl 3-Oxo-3-(o-tolyl)propionate, 3-Oxo-3-(o-tolyl)propionic Acid Ethyl Ester
PubChem CID 2760276
IUPAC Name ethyl 3-(2-methylphenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 206.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl (2-Methylbenzoyl)acetate
EINECSNumber (3)-1762
TSCA No