missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Methyl-3-furancarboxylate 96.0+%, TCI America™

Catalog No. M114010G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M114010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M114010G Supplier TCI America Supplier No. M114010G
Only null left

Chemical Identifiers

CAS 28921-35-9
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00134327
InChI Key DOHUEYINVDLUSG-UHFFFAOYSA-N
Synonym 2-Methyl-3-furancarboxylic Acid Ethyl Ester, Ethyl 2-Methyl-3-furoate, 2-Methyl-3-furoic Acid Ethyl Ester
PubChem CID 261900
IUPAC Name ethyl 2-methylfuran-3-carboxylate
SMILES CCOC(=O)C1=C(C)OC=C1

Specifications

Color Yellow
Boiling Point 87°C
Quantity 10 g
Formula Weight 154.17
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Methyl-3-furancarboxylate