missing translation for 'onlineSavingsMsg'
Learn More

Ethyl(2-methoxyethyl)dimethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. e12815g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12815G Supplier TCI America Supplier No. E12815G
Only null left

Chemical Identifiers

CAS 557788-37-1
Molecular Formula C9H18F6N2O5S2
Molecular Weight (g/mol) 412.362
MDL Number MFCD09038873
InChI Key NGLLWWMHAWYWLY-UHFFFAOYSA-N
PubChem CID 87081346
IUPAC Name bis(trifluoromethylsulfonyl)azanide;ethyl-(2-methoxyethyl)-dimethylazanium
SMILES CC[N+](C)(C)CCOC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 412.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl(2-methoxyethyl)dimethylammonium Bis(trifluoromethanesulfonyl)imide