missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Imidazolecarboxylate 98.0+%, TCI America™

Catalog No. p-7129243
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
E09521G 1 g
E09525G 5 g
2 options

Catalog No. E09521G

Supplier: TCI America E09521G

Only null left

Chemical Identifiers

CAS 33543-78-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD03426031
InChI Key UHYNYIGCGVDBTC-UHFFFAOYSA-N
Synonym ethyl imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid, ethyl ester, ethyl1h-imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid ethyl ester, ethyl 2-imidazolecarboxylate, imidazole-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-1h-imidazole, ethylimidazole-2-carboxylate, ethoxyformyl imidazole, zlchem 847
PubChem CID 549404
IUPAC Name ethyl 1H-imidazole-2-carboxylate
SMILES CCOC(=O)C1=NC=CN1

Specifications

Melting Point 181°C
Color White-Yellow
Boiling Point 51°C
Quantity 1 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Imidazolecarboxylate