missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Hydroxybutyrate 95.0+%, TCI America™
$481.30 - $1440.03
Chemical Identifiers
| CAS | 52089-54-0 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.16 |
| MDL Number | MFCD00014407 |
| InChI Key | KWWOQRSLYPHAMK-UHFFFAOYNA-N |
| Synonym | 2-Hydroxybutyric Acid Ethyl Ester |
| PubChem CID | 521365 |
| IUPAC Name | ethyl 2-hydroxybutanoate |
| SMILES | CCOC(=O)C(O)CC |
Chemical Identifiers
| 52089-54-0 | |
| 132.16 | |
| KWWOQRSLYPHAMK-UHFFFAOYNA-N | |
| 521365 | |
| CCOC(=O)C(O)CC |
| C6H12O3 | |
| MFCD00014407 | |
| 2-Hydroxybutyric Acid Ethyl Ester | |
| ethyl 2-hydroxybutanoate |
Specifications
| 52089-54-0 | |
| 167°C | |
| MFCD00014407 | |
| 2-Hydroxybutyric Acid Ethyl Ester | |
| CCOC(=O)C(O)CC | |
| 132.16 | |
| 132.16 | |
| Liquid |
| Yellow | |
| C6H12O3 | |
| 5 g | |
| KWWOQRSLYPHAMK-UHFFFAOYNA-N | |
| ethyl 2-hydroxybutanoate | |
| 521365 | |
| ≥95.0% (GC) | |
| Ethyl 2-Hydroxybutyrate |
Safety and Handling
EINECSNumber : (2)-1430
TSCA : No