missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Hydroxybutyrate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 167°C |
| Quantity | 5 g |
| Formula Weight | 132.16 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl 2-Hydroxybutyrate |
Safety and Handling
| EINECSNumber | (2)-1430 |
| TSCA | No |