missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Hydroxy-3-butynoate 95.0+%, TCI America™

Catalog No. H08231G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H08231G Supplier TCI America Supplier No. H08231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18418-08-1
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.127
MDL Number MFCD00191654
InChI Key GDWLVFKOPXSIAE-UHFFFAOYSA-N
Synonym 2-Hydroxy-3-butynoic Acid Ethyl Ester
PubChem CID 44123461
IUPAC Name ethyl 2-hydroxybut-3-ynoate
SMILES CCOC(=O)C(C#C)O

Specifications

Color Yellow
Boiling Point 78°C
Quantity 1 g
Formula Weight 128.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Hydroxy-3-butynoate
TSCA No
Recommended Storage Freezer