missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-furoate, 99%

CAS: 614-99-3 | C7H8O3 | 140.138 g/mol

Supplier:  Thermo Scientific Chemicals A1390922

 View more versions of this product

Catalog No. AAA1390922

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 2-furoate
31°C to 36°C
195°C to 196°C
C7H8O3
100 g
14,4307
NHXSTXWKZVAVOQ-UHFFFAOYSA-N
ethyl furan-2-carboxylate
11980
99%
614-99-3
1.12
70°C (158°F)
MFCD00003237
2653
ethyl 2-furoate, ethyl furoate, ethyl 2-furancarboxylate, 2-furancarboxylic acid, ethyl ester, ethyl pyromucate, 2-carboethoxyfuran, furan-2-carboxylic acid ethyl ester, ethyl-2-furoate, 2-furoic acid, ethyl ester, ethyl2-furoate
CCOC(=O)C1=CC=CO1
140.138
140.14

Chemical Identifiers

614-99-3
140.138
NHXSTXWKZVAVOQ-UHFFFAOYSA-N
ethyl furan-2-carboxylate
C7H8O3
MFCD00003237
11980
CCOC(=O)C1=CC=CO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-404-0

RTECSNumber : LV1850000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only