missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-furoate, 99%

Catalog No. AAA1390914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1390914 25 g
AAA1390922 100 g
AAA1390936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1390914 Supplier Thermo Scientific Chemicals Supplier No. A1390914

CAS: 614-99-3 | C7H8O3 | 140.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-99-3
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003237
InChI Key NHXSTXWKZVAVOQ-UHFFFAOYSA-N
PubChem CID 11980
IUPAC Name ethyl furan-2-carboxylate
SMILES CCOC(=O)C1=CC=CO1

Specifications

Melting Point 31°C to 36°C
Density 1.12
Boiling Point 195°C to 196°C
Flash Point 70°C (158°F)
Quantity 25 g
Beilstein 2653
Merck Index 14,4307
Synonym ethyl 2-furoate, ethyl furoate, ethyl 2-furancarboxylate, 2-furancarboxylic acid, ethyl ester, ethyl pyromucate, 2-carboethoxyfuran, furan-2-carboxylic acid ethyl ester, ethyl-2-furoate, 2-furoic acid, ethyl ester, ethyl2-furoate
Formula Weight 140.14
Percent Purity 99%
Chemical Name or Material Ethyl 2-furoate
Show More Show Less
EINECSNumber 210-404-0
RTECSNumber LV1850000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only