missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Furancarboxylate 98.0+%, TCI America™

Catalog No. F009825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F009825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F009825G Supplier TCI America Supplier No. F009825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 614-99-3
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003237
InChI Key NHXSTXWKZVAVOQ-UHFFFAOYSA-N
Synonym ethyl 2-furoate, ethyl furoate, ethyl 2-furancarboxylate, 2-furancarboxylic acid, ethyl ester, ethyl pyromucate, 2-carboethoxyfuran, furan-2-carboxylic acid ethyl ester, ethyl-2-furoate, 2-furoic acid, ethyl ester, ethyl2-furoate
PubChem CID 11980
IUPAC Name ethyl furan-2-carboxylate
SMILES CCOC(=O)C1=CC=CO1

Specifications

Boiling Point 196°C
Quantity 25 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 2-Furancarboxylate
EINECSNumber (5)-0046
RTECSNumber LV1850000
TSCA Yes
Recommended Storage Refrigerator