missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Fluoropropionate 97.0+%, TCI America™

Catalog No. E06635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06635G 5 g
E066325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06635G Supplier TCI America Supplier No. E06635G

Chemical Identifiers

CAS 349-43-9
Molecular Formula C5H9FO2
Molecular Weight (g/mol) 120.12
MDL Number MFCD01320801
InChI Key ODMITNOQNBVSQG-UHFFFAOYNA-N
Synonym 2-Fluoropropionic Acid Ethyl Ester
PubChem CID 265947
IUPAC Name ethyl 2-fluoropropanoate
SMILES CCOC(=O)C(C)F

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 g
UN Number 3272
Formula Weight 120.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Fluoropropionate