missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Ethylbutyrate 99.0+%, TCI America™
$167.76 - $1357.03
Chemical Identifiers
| CAS | 2983-38-2 |
|---|---|
| Molecular Formula | C8H16O2 |
| Molecular Weight (g/mol) | 144.214 |
| MDL Number | MFCD00059396 |
| InChI Key | MJGSLNIPTRPYJV-UHFFFAOYSA-N |
| Synonym | ethyl 2-ethylbutyrate, butanoic acid, 2-ethyl-, ethyl ester, 2-ethyl-n-butyric acid ethyl ester, unii-9zb2g5ff44, ethyl2-ethylbutyrate, ethyl 2-ethyl butyrate, acmc-209hbb, 2-ethylbutyric acid ethyl ester, mjgslniptrpyjv-uhfffaoysa |
| PubChem CID | 76326 |
| IUPAC Name | ethyl 2-ethylbutanoate |
| SMILES | CCC(CC)C(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E034525ML
|
TCI America
E034525ML |
25 mL |
Each for $167.76
|
|
|||||
|
E0345500ML
|
TCI America
E0345500ML |
500 mL |
Each for $1,357.03
|
|
|||||
Chemical Identifiers
| 2983-38-2 | |
| 144.214 | |
| MJGSLNIPTRPYJV-UHFFFAOYSA-N | |
| 76326 | |
| CCC(CC)C(=O)OCC |
| C8H16O2 | |
| MFCD00059396 | |
| ethyl 2-ethylbutyrate, butanoic acid, 2-ethyl-, ethyl ester, 2-ethyl-n-butyric acid ethyl ester, unii-9zb2g5ff44, ethyl2-ethylbutyrate, ethyl 2-ethyl butyrate, acmc-209hbb, 2-ethylbutyric acid ethyl ester, mjgslniptrpyjv-uhfffaoysa | |
| ethyl 2-ethylbutanoate |
Specifications
| 2983-38-2 | |
| 152°C | |
| MFCD00059396 | |
| 3272 | |
| MJGSLNIPTRPYJV-UHFFFAOYSA-N | |
| ethyl 2-ethylbutanoate | |
| 76326 | |
| ≥99.0% (GC) | |
| Ethyl 2-Ethylbutyrate |
| Yellow | |
| C8H16O2 | |
| 25 mL | |
| ethyl 2-ethylbutyrate, butanoic acid, 2-ethyl-, ethyl ester, 2-ethyl-n-butyric acid ethyl ester, unii-9zb2g5ff44, ethyl2-ethylbutyrate, ethyl 2-ethyl butyrate, acmc-209hbb, 2-ethylbutyric acid ethyl ester, mjgslniptrpyjv-uhfffaoysa | |
| CCC(CC)C(=O)OCC | |
| 144.214 | |
| 144.21 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0782
TSCA : No