missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Ethylbutyrate 99.0+%, TCI America™

Catalog No. E034525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E034525ML 25 mL
E0345500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E034525ML Supplier TCI America Supplier No. E034525ML
Only null left

Chemical Identifiers

CAS 2983-38-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00059396
InChI Key MJGSLNIPTRPYJV-UHFFFAOYSA-N
Synonym ethyl 2-ethylbutyrate, butanoic acid, 2-ethyl-, ethyl ester, 2-ethyl-n-butyric acid ethyl ester, unii-9zb2g5ff44, ethyl2-ethylbutyrate, ethyl 2-ethyl butyrate, acmc-209hbb, 2-ethylbutyric acid ethyl ester, mjgslniptrpyjv-uhfffaoysa
PubChem CID 76326
IUPAC Name ethyl 2-ethylbutanoate
SMILES CCC(CC)C(=O)OCC

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Ethylbutyrate
EINECSNumber (2)-0782
TSCA No