missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Ethylacetoacetate 97.0+%, TCI America™

Catalog No. E089525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E089525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E089525G Supplier TCI America Supplier No. E089525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 607-97-6
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00039898
InChI Key OKANYBNORCUPKZ-UHFFFAOYSA-N
Synonym ethyl 2-ethylacetoacetate, ethyl 2-acetylbutyrate, butanoic acid, 2-ethyl-3-oxo-, ethyl ester, 2-ethylacetoacetic acid ethyl ester, ethyl 2-ethyl-3-ketobutyrate, ethyl alpha-ethylacetoacetate, ethyl-2-ethylacetoacetate, 2-ethyl-3-oxo-butyric acid ethyl ester, ethyl .alpha.-acetylbutyrate, ethyl 2-ethyl-3-oxo-butanoate
PubChem CID 79065
IUPAC Name ethyl 2-ethyl-3-oxobutanoate
SMILES CCC(C(=O)C)C(=O)OCC

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 g
Formula Weight 158.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Ethylacetoacetate
EINECSNumber (2)-1505
TSCA Yes