missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-ethylacetoacetate, 95%

Catalog No. p-7026340
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2229514 25 g
AAB2229522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2229514 Supplier Thermo Scientific Chemicals Supplier No. B2229514

CAS: 607-97-6 | C8H14O3 | 158.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 607-97-6
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00039898
InChI Key OKANYBNORCUPKZ-UHFFFAOYSA-N
Synonym ethyl 2-ethylacetoacetate, ethyl 2-acetylbutyrate, butanoic acid, 2-ethyl-3-oxo-, ethyl ester, 2-ethylacetoacetic acid ethyl ester, ethyl 2-ethyl-3-ketobutyrate, ethyl alpha-ethylacetoacetate, ethyl-2-ethylacetoacetate, 2-ethyl-3-oxo-butyric acid ethyl ester, ethyl .alpha.-acetylbutyrate, ethyl 2-ethyl-3-oxo-butanoate
PubChem CID 79065
IUPAC Name ethyl 2-ethyl-3-oxobutanoate
SMILES CCC(C(=O)C)C(=O)OCC

Specifications

Density 1.009
Boiling Point 83°C to 85°C (10 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.422
Quantity 25 g
Beilstein 636286
Formula Weight 158.2
Percent Purity 95%
Chemical Name or Material Ethyl 2-ethylacetoacetate
Hazard Category Warning
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-151-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only