missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 96.0+%, TCI America™

Catalog No. p-7129323
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10875G 5 g
E108725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10875G Supplier TCI America Supplier No. E10875G
Only null left

Chemical Identifiers

CAS 571-55-1
Molecular Formula C9H11F3O4
Molecular Weight (g/mol) 240.178
MDL Number MFCD02677683
InChI Key XNGGOXOLHQANRB-AATRIKPKSA-N
Synonym ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, e-ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2-ethoxymethylene-4,4,4-trifluoroacetoacetate, ethyl 2e-2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoate, ethyl ethoxymethylene-3-oxo-4,4,4-trifluoro-butyrate, ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2e-3-ethoxy-2-2,2,2-trifluoroacetyl prop-2-enoate, 2e-2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoic acid ethyl ester
PubChem CID 6185754
IUPAC Name ethyl (2E)-2-(ethoxymethylidene)-4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC=C(C(=O)C(F)(F)F)C(=O)OCC

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 240.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate
TSCA No
Recommended Storage Refrigerator