missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, 97%

Catalog No. AAH2592709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2592709 10 g
AAH2592718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2592709 Supplier Thermo Scientific Chemicals Supplier No. H2592709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 571-55-1 | C9H11F3O4 | 240.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 571-55-1
Molecular Formula C9H11F3O4
Molecular Weight (g/mol) 240.178
MDL Number MFCD02677683
InChI Key XNGGOXOLHQANRB-AATRIKPKSA-N
Synonym ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate, e-ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2-ethoxymethylene-4,4,4-trifluoroacetoacetate, ethyl 2e-2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoate, ethyl ethoxymethylene-3-oxo-4,4,4-trifluoro-butyrate, ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutanoate, ethyl 2e-3-ethoxy-2-2,2,2-trifluoroacetyl prop-2-enoate, 2e-2-ethoxymethylidene-4,4,4-trifluoro-3-oxobutanoic acid ethyl ester
PubChem CID 6185754
IUPAC Name ethyl (2E)-2-(ethoxymethylidene)-4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC=C(C(=O)C(F)(F)F)C(=O)OCC

Specifications

Density 1.235
Boiling Point 80°C to 82°C (1 mmHg)
Flash Point 104°C (220°F)
Refractive Index 1.429
Quantity 10 g
Formula Weight 240.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-ethoxymethylene-4,4,4-trifluoro-3-oxobutyrate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only