missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(Ethoxymethylene)-2-cyanoacetate 98.0+%, TCI America™

Catalog No. p-7128949
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E034125G 25 g
E0341500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E034125G Supplier TCI America Supplier No. E034125G

Chemical Identifiers

CAS 94-05-3
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00009136
InChI Key KTMGNAIGXYODKQ-SREVYHEPSA-N
Synonym ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
PubChem CID 1715183
IUPAC Name ethyl (2Z)-2-cyano-3-ethoxyprop-2-enoate
SMILES CCO\C=C(\C#N)C(=O)OCC

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 191°C
Quantity 25 g
UN Number 3439
Formula Weight 169.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-(Ethoxymethylene)-2-cyanoacetate
EINECSNumber (9)-0242
TSCA Yes