missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Cyano-2-methylpropionate 98.0+%, TCI America™
Supplier: TCI America C154625G
Specifications
| Ethyl 2-Cyano-2-methylpropionate | |
| Colorless | |
| C7H11NO2 | |
| 25 g | |
| FYGRPGOHQCPZCV-UHFFFAOYSA-N | |
| ethyl 2-cyano-2-methylpropanoate | |
| 344887 | |
| ≥98.0% (GC) |
| 1572-98-1 | |
| 185°C | |
| MFCD00034667 | |
| 2-cyano-2-methylpropionic acid ethyl ester, ethyl 2-cyano-2-methylpropionate, ethyl 2-cyano-2,2-dimethylacetate, 2-cyano-2-methylpropanoic acid ethyl ester, ethyl 2-cyano-2-methyl-propanoate, propanoic acid, 2-cyano-2-methyl-, ethyl ester, pubchem23352, acmc-209dfv, ksc529g3h, ethyl2-cyano-2-methylpropionate | |
| CCOC(=O)C(C)(C)C#N | |
| 141.17 | |
| 141.17 | |
| Liquid |
Chemical Identifiers
| 1572-98-1 | |
| 141.17 | |
| FYGRPGOHQCPZCV-UHFFFAOYSA-N | |
| ethyl 2-cyano-2-methylpropanoate |
| C7H11NO2 | |
| MFCD00034667 | |
| 344887 | |
| CCOC(=O)C(C)(C)C#N |
Safety and Handling
TSCA : No