missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-cyano-2-methylpropionate, 97%

Catalog No. AAH3143209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3143209 10 g
AAH3143218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3143209 Supplier Thermo Scientific Chemicals Supplier No. H3143209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1572-98-1 | C7H11NO2 | 141.17 g/mol

It is employed as a intermediates for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediates for pharmaceutical and organic synthesis.

Solubility
Soluble in ether. Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1572-98-1
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00034667
InChI Key FYGRPGOHQCPZCV-UHFFFAOYSA-N
PubChem CID 344887
IUPAC Name ethyl 2-cyano-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)C#N

Specifications

Density 0.97
Boiling Point 185°C
Quantity 10 g
Synonym 2-cyano-2-methylpropionic acid ethyl ester, ethyl 2-cyano-2-methylpropionate, ethyl 2-cyano-2,2-dimethylacetate, 2-cyano-2-methylpropanoic acid ethyl ester, ethyl 2-cyano-2-methyl-propanoate, propanoic acid, 2-cyano-2-methyl-, ethyl ester, pubchem23352, acmc-209dfv, ksc529g3h, ethyl2-cyano-2-methylpropionate
Solubility Information Soluble in ether. Not miscible or difficult to mix with water.
Formula Weight 141.17
Percent Purity 97%
Chemical Name or Material Ethyl 2-cyano-2-methylpropionate
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only