missing translation for 'onlineSavingsMsg'
Learn More

Éthyle 2-chloronicotinate 98,0+%, TCI America™

Numéro de catalogue. p-7129181
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E084425G 25 g
E0844100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E084425G Fournisseur TCI America Code fournisseur E084425G

Identifiants chimiques

CAS 1452-94-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD00674129
InChI Key PMIMPBYTPPRBGD-UHFFFAOYSA-N
Synonym ethyl 2-chloronicotinate, 2-chloronicotinic acid ethyl ester, ethyl-2-chloronicotinate, 2-chloro-nicotinic acid ethyl ester, ethyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-3-pyridinecarboxylic acid ethyl ester, ethyl 2chloronicotinate, acmc-1brbe, ethyl 2-chloro-nicotinate
PubChem CID 2796614
IUPAC Name Éthyle 2-chloropyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)Cl

Spécifications

Color Jaune
Boiling Point 78°C
Quantity 25 g
Formula Weight 185.61
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material Ethyl 2-Chloronicotinate