missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Butylacetoacetate 96.0+%, TCI America™

Catalog No. B07025ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B07025ML 5 mL
B070225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B07025ML Supplier TCI America Supplier No. B07025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1540-29-0
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD00026888
InChI Key ZTOQBHVLCJERBS-UHFFFAOYSA-N
Synonym ethyl 2-n-butylacetoacetate, ethyl 2-butylacetoacetate, hexanoic acid, 2-acetyl-, ethyl ester, ethyl .alpha.-butylacetoacetate, 2-acetylhexanoic acid ethyl ester, ethyl butyl acetoacetate, ethyl 2-acetyl hexanoate, acmc-209da5, ethyl-2-n-butylacetoacetate, ksc494i1n
PubChem CID 95466
IUPAC Name ethyl 2-acetylhexanoate
SMILES CCCCC(C(=O)C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 224°C
Quantity 5 mL
Formula Weight 186.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Butylacetoacetate
EINECSNumber (2)-1505
TSCA No