missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromovalerate 98.0+%, TCI America™

Catalog No. p-7129254
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
Catalog No. Quantity
E097025G 25 g
E0970100G 100 g
2 options

Catalog No. E097025G

Supplier: TCI America E097025G

Only null left

Chemical Identifiers

CAS 615-83-8
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00000159
InChI Key ORSIRXYHFPHWTN-UHFFFAOYSA-N
PubChem CID 79071
IUPAC Name ethyl 2-bromopentanoate
SMILES CCCC(C(=O)OCC)Br

Specifications

Color Yellow
Boiling Point 94°C
Quantity 25 g
UN Number 3265
Synonym ethyl 2-bromovalerate, ethyl2-bromovalerate, ethyl .alpha.-bromovalerate, pentanoic acid, 2-bromo-, ethyl ester, 2-bromovaleric acid ethyl ester, 2-bromopentanoic acid ethyl ester, 2-bromo-pentanoic acid ethyl ester, pentanoic acid, bromo-, ethyl ester, ethyl bromovalerate, ethyl-2bromovalerate
Formula Weight 209.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromovalerate