missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromovalerate, 97%

Catalog No. p-7028294
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2553014 25 g
AAB2553022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2553014 Supplier Thermo Scientific Chemicals Supplier No. B2553014
Only null left

CAS: 615-83-8 | C7H13BrO2 | 209.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-83-8
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00000159
InChI Key ORSIRXYHFPHWTN-UHFFFAOYSA-N
PubChem CID 79071
IUPAC Name ethyl 2-bromopentanoate
SMILES CCCC(C(=O)OCC)Br

Specifications

Density 1.226
Boiling Point 190°C to 192°C
Flash Point 77°C (170°F)
Refractive Index 1.448
Quantity 25 g
UN Number UN3265
Beilstein 1098901
Synonym ethyl 2-bromovalerate, ethyl2-bromovalerate, ethyl .alpha.-bromovalerate, pentanoic acid, 2-bromo-, ethyl ester, 2-bromovaleric acid ethyl ester, 2-bromopentanoic acid ethyl ester, 2-bromo-pentanoic acid ethyl ester, pentanoic acid, bromo-, ethyl ester, ethyl bromovalerate, ethyl-2bromovalerate
Formula Weight 209.09
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromovalerate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-450-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only