missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromothiazole-5-carboxylate 98.0+%, TCI America™

Catalog No. p-7129205
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
E08911G 1 g
E08915G 5 g
2 options

Catalog No. E08911G

Supplier: TCI America E08911G

Only null left

Chemical Identifiers

CAS 41731-83-3
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.08
MDL Number MFCD00463837
InChI Key KTYIFXLNIMPSKI-UHFFFAOYSA-N
PubChem CID 3614103
IUPAC Name ethyl 2-bromo-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=CN=C(Br)S1

Specifications

Color Yellow
Boiling Point 68°C
Quantity 1 g
Synonym ethyl 2-bromothiazole-5-carboxylate, 2-bromo-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-5-thiazolecarboxylate, 2-bromothiazole-5-carboxylic acid ethyl ester, 2-bromo-5-ethoxycarbonyl thiazole, ethyl-2-bromothiazole-5-carboxylate, ethyl2-bromo-5-thiazolecarboxylate, ethyl2-bromothiazole-5-carboxylate, pubchem13480, acmc-1ambg
Formula Weight 236.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromothiazole-5-carboxylate