missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromothiazole-5-carboxylate, 98%

Catalog No. p-7036751
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5510903 1 g
AAH5510906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5510903 Supplier Thermo Scientific Chemicals Supplier No. H5510903
Only null left

CAS: 41731-83-3 | C6H6BrNO2S | 236.08 g/mol

Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromothiazole-5-carboxylate is used in medicine, pharmaceutical intermediate

Solubility
Miscible is water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 41731-83-3
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.08
MDL Number MFCD00463837
InChI Key KTYIFXLNIMPSKI-UHFFFAOYSA-N
PubChem CID 3614103
IUPAC Name ethyl 2-bromo-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=CN=C(Br)S1

Specifications

Density 1.581
Boiling Point 216°C to 217°C
Flash Point 110°C (230°F)
Refractive Index 1.553
Quantity 1 g
Synonym ethyl 2-bromothiazole-5-carboxylate, 2-bromo-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-5-thiazolecarboxylate, 2-bromothiazole-5-carboxylic acid ethyl ester, 2-bromo-5-ethoxycarbonyl thiazole, ethyl-2-bromothiazole-5-carboxylate, ethyl2-bromo-5-thiazolecarboxylate, ethyl2-bromothiazole-5-carboxylate, pubchem13480, acmc-1ambg
Solubility Information Miscible is water.
Formula Weight 236.08
Percent Purity 98%
Chemical Name or Material Ethyl 2-bromothiazole-5-carboxylate
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only