missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromopropionate 98.0+%, TCI America™

Catalog No. p-7118587
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B064625G 25 g
B0646100G 100 g
B0646500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B064625G Supplier TCI America Supplier No. B064625G
Only null left

Chemical Identifiers

CAS 535-11-5
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000144
InChI Key ARFLASKVLJTEJD-UHFFFAOYNA-N
Synonym ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate
PubChem CID 79040
IUPAC Name ethyl 2-bromopropanoate
SMILES CCOC(=O)C(C)Br

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 1992
Formula Weight 181.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromopropionate
EINECSNumber (2)-1179&(2)-2647
RTECSNumber UA2451730
TSCA Yes