missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromopropionate, 98+%

Catalog No. AAA1082018
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1082018 50 g
AAA1082030 250 g
AAA1082036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1082018 Supplier Thermo Scientific Chemicals Supplier No. A1082018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol

It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

Solubility
Immiscible in water. Soluble in alcohol and ether.

Notes
Store in dark and away from oxidizing agents and acids.

Chemical Identifiers

CAS 535-11-5
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000144
InChI Key ARFLASKVLJTEJD-UHFFFAOYNA-N
Synonym ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate
PubChem CID 79040
IUPAC Name ethyl 2-bromopropanoate
SMILES CCOC(=O)C(C)Br

Specifications

Density 1.391
Boiling Point 158°C to 160°C
Flash Point 51°C (123°F)
Refractive Index 1.446
Quantity 50 g
UN Number UN1992
Beilstein 773920
Merck Index 14,3772
Solubility Information Immiscible in water. Soluble in alcohol and ether.
Formula Weight 181.03
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromopropionate
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H318-H226-H302-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 208-609-5
RTECSNumber UA2451730
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only