missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromonicotinate, 96%

Catalog No. AAH52335MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52335MD 250 mg
AAH5233503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52335MD Supplier Thermo Scientific Chemicals Supplier No. H52335MD
Only null left

CAS: 53087-78-8 | C8H8BrNO2 | 230.061 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53087-78-8
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD04113825
InChI Key QOFKYVXBHUEWBX-UHFFFAOYSA-N
Synonym ethyl 2-bromonicotinate, 2-bromonicotinic acid ethyl ester, 2-bromonicotinic ethyl ester, ethyl 2-bromonicotite, pubchem12886, ethyl 2-bromanylpyridine-3-carboxylate, 2-bromo-nicotinic acid ethyl ester, ethyl 2-bromo-3-pyridinecarboxylate, 2-bromo-3-pyridinecarboxylic acid ethyl ester, 2-bromo-pyridine-3-carboxylic acid ethyl ester
PubChem CID 1415877
IUPAC Name ethyl 2-bromopyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)Br

Specifications

Quantity 250 mg
Formula Weight 230.07
Percent Purity 96%
Chemical Name or Material Ethyl 2-bromonicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only