missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromobutyrate 98.0+%, TCI America™

Catalog No. p-7118678
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B080025G 25 g
B0800500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B080025G Supplier TCI America Supplier No. B080025G
Only null left

Chemical Identifiers

CAS 533-68-6
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000155
InChI Key XIMFCGSNSKXPBO-UHFFFAOYNA-N
Synonym ethyl 2-bromobutyrate, dl-ethyl 2-bromobutyrate, 2-bromobutanoic acid ethyl ester, butanoic acid, 2-bromo-, ethyl ester, 2-bromobutyric acid ethyl ester, ethyl .alpha.-bromobutyrate, 2-bromo-butyric acid ethyl ester, butyric acid, 2-bromo-, ethyl ester, 2-bromo-butanoicaciethylester, ethyl alpha-bromobutyrate
PubChem CID 79039
IUPAC Name ethyl 2-bromobutanoate
SMILES CCOC(=O)C(Br)CC

Specifications

Color Yellow
Boiling Point 178°C
Quantity 25 g
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromobutyrate
EINECSNumber (2)-1179
TSCA Yes