missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Bromobutyrate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 178°C |
| Quantity | 25 g |
| Formula Weight | 195.06 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl 2-Bromobutyrate |
Safety and Handling
| EINECSNumber | (2)-1179 |
| TSCA | Yes |