Learn More
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate, 95%, Thermo Scientific Chemicals
$617.86 - $617.86
Chemical Identifiers
| CAS | 238749-50-3 |
|---|---|
| Molecular Formula | C9H8BrNO2S |
| Molecular Weight (g/mol) | 274.132 |
| MDL Number | MFCD15730638 |
| InChI Key | KXGSKTSSCQBDOA-UHFFFAOYSA-N |
| Synonym | ethyl 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylate, ethyl 2-bromothieno<3,2-b>pyrrole-5-carboxylate, 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4h-thieno 3.2-b pyrrole-5-carboxylate, 2-bromothieno 3,2-b-pyrrole-5-carboxylic acid ethyl ester, 2-bromothieno 3,2-b pyrrole-5-carboxylic acid ethyl ester |
| PubChem CID | 13164349 |
| IUPAC Name | ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate |
| SMILES | CCOC(=O)C1=CC2=C(N1)C=C(S2)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5907603
|
Thermo Scientific Chemicals
H5907603 |
1 g |
Each for $617.86
|
|
|||||
Description
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.
Solubility
Sparingly soluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 238749-50-3 | |
| 274.132 | |
| KXGSKTSSCQBDOA-UHFFFAOYSA-N | |
| 13164349 | |
| CCOC(=O)C1=CC2=C(N1)C=C(S2)Br |
| C9H8BrNO2S | |
| MFCD15730638 | |
| ethyl 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylate, ethyl 2-bromothieno<3,2-b>pyrrole-5-carboxylate, 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4h-thieno 3.2-b pyrrole-5-carboxylate, 2-bromothieno 3,2-b-pyrrole-5-carboxylic acid ethyl ester, 2-bromothieno 3,2-b pyrrole-5-carboxylic acid ethyl ester | |
| ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate |
Specifications
| 238749-50-3 | |
| MFCD15730638 | |
| 4426004 | |
| Sparingly soluble in water. | |
| CCOC(=O)C1=CC2=C(N1)C=C(S2)Br | |
| 274.132 | |
| 274.13 | |
| Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate |
| C9H8BrNO2S | |
| 1 g | |
| ethyl 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylate, ethyl 2-bromothieno<3,2-b>pyrrole-5-carboxylate, 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4h-thieno 3.2-b pyrrole-5-carboxylate, 2-bromothieno 3,2-b-pyrrole-5-carboxylic acid ethyl ester, 2-bromothieno 3,2-b pyrrole-5-carboxylic acid ethyl ester | |
| KXGSKTSSCQBDOA-UHFFFAOYSA-N | |
| ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate | |
| 13164349 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only