missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromo-3-methylbutyrate, 95%, Thermo Scientific™

Catalog No. AC409790100
Change view
Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409790100 10g Glass bottle
AC409790500 50g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC409790100 Supplier Thermo Scientific Chemicals Supplier No. 409790100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 609-12-1
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
InChI Key WNFUWONOILPKNX-UHFFFAOYSA-N
Synonym ethyl 2-bromoisovalerate, ethyl 2-bromo-3-methylbutyrate, butanoic acid, 2-bromo-3-methyl-, ethyl ester, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, butyric acid, 2-bromo-3-methyl-, ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, ethyl 2-bromo isovalerate
PubChem CID 79068
IUPAC Name ethyl 2-bromo-3-methylbutanoate
SMILES CCOC(=O)C(C(C)C)Br

Specifications

Boiling Point 77.0°C (12.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4485 to 1.4505
Linear Formula (CH3)2CHCHBrCOOC2H5
Quantity 10g
Formula Weight 209.08
Percent Purity 95%
Chemical Name or Material Ethyl 2-bromo-3-methylbutyrate
Show More Show Less
EINECSNumber 210-178-3

RUO – Research Use Only