missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Benzoylpropionate 98.0+%, TCI America™

Catalog No. p-7129313
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10711G 1 g
E10715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10711G Supplier TCI America Supplier No. E10711G
Only null left

Chemical Identifiers

CAS 10488-87-6
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00026882
InChI Key RAWGHPXIJSCHFZ-UHFFFAOYNA-N
Synonym 2-Benzoylpropionic Acid Ethyl Ester
PubChem CID 139166
IUPAC Name ethyl 2-methyl-3-oxo-3-phenylpropanoate
SMILES CCOC(=O)C(C)C(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 96°C
Quantity 1 g
Formula Weight 206.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Benzoylpropionate