missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Aminoisonicotinate 98.0+%, TCI America™

Catalog No. E09441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09441G 1 g
E09445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09441G Supplier TCI America Supplier No. E09441G

Chemical Identifiers

CAS 13362-30-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD03791260
InChI Key XVBZFXZNJAFCHL-UHFFFAOYSA-N
Synonym ethyl 2-aminoisonicotinate, 2-amino-isonicotinic acid ethyl ester, ethyl 2-amino-4-pyridinecarboxylate, 2-aminoisonicotinic acid ethyl ester, 2-amino-4-ethoxycarbonyl pyridine, 4-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino-4-pyridinecarboxylic acid ethyl ester, 2-aminopyridine-4-carboxylic acid ethyl ester, pubchem16599, acmc-209bt6
PubChem CID 7015521
IUPAC Name ethyl 2-aminopyridine-4-carboxylate
SMILES CCOC(=O)C1=CC(=NC=C1)N

Specifications

Melting Point 125°C
Color Yellow
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Aminoisonicotinate