missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Aminobenzothiazole-6-carboxylate 98.0+%, TCI America™

Catalog No. p-7129266
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09851G 1 g
E09855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09851G Supplier TCI America Supplier No. E09851G
Only null left

Chemical Identifiers

CAS 50850-93-6
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.262
MDL Number MFCD00102724
InChI Key VYJSGJXWKSDUSG-UHFFFAOYSA-N
Synonym ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, ethyl2-amino-benzothiazole-6-carboxylate, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 2-amino benzo thiazole-6-carboxylic acid ethyl ester, 3kid
PubChem CID 601008
IUPAC Name ethyl 2-amino-1,3-benzothiazole-6-carboxylate
SMILES CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N

Specifications

Melting Point 242°C
Color Yellow
Quantity 1 g
Formula Weight 222.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Aminobenzothiazole-6-carboxylate