missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Aminobenzothiazole-6-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 242°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 222.26 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ethyl 2-Aminobenzothiazole-6-carboxylate |
Safety and Handling
| TSCA | No |