missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-5-methylthiophene-3-carboxylate, 95%

Catalog No. AAH6176606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6176606 5 g
AAH6176603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6176606 Supplier Thermo Scientific Chemicals Supplier No. H6176606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4815-32-1 | C8H11NO2S | 185.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4815-32-1
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD01922142
InChI Key AYBUNZGJQVYGTM-UHFFFAOYSA-N
Synonym 2-amino-5-methyl-thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methylthiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-5-methyl-, ethyl ester, ethyl2-amino-5-methyl-3-thiophenecarboxylate, 2-amino-5-methyl-3-thiophenecarboxylic acid ethyl ester, ethyl 2-amino-5-methyl-thiophene-3-carboxylate, ksc590m8t, aronis25101, 2-amino-5-methyl-thiophene-3-carboxylicacidethylester
PubChem CID 4062885
IUPAC Name ethyl 2-amino-5-methylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1)C)N

Specifications

Quantity 5 g
Formula Weight 185.24
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 2-amino-5-methylthiophene-3-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only