missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (2-Amino-4-thiazolyl)acetate 98.0+%, TCI America™
$44.73 - $333.67
Chemical Identifiers
| CAS | 53266-94-7 |
|---|---|
| Molecular Formula | C7H10N2O2S |
| Molecular Weight (g/mol) | 186.23 |
| MDL Number | MFCD00005330 |
| InChI Key | SHQNGLYXRFCPGZ-UHFFFAOYSA-N |
| Synonym | ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate |
| PubChem CID | 104454 |
| IUPAC Name | ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate |
| SMILES | CCOC(=O)CC1=CSC(N)=N1 |
Chemical Identifiers
| 53266-94-7 | |
| 186.23 | |
| SHQNGLYXRFCPGZ-UHFFFAOYSA-N | |
| 104454 | |
| CCOC(=O)CC1=CSC(N)=N1 |
| C7H10N2O2S | |
| MFCD00005330 | |
| ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate | |
| ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate |
Specifications
| 53266-94-7 | |
| C7H10N2O2S | |
| 25 g | |
| SHQNGLYXRFCPGZ-UHFFFAOYSA-N | |
| ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate | |
| 104454 | |
| ≥98.0% (GC,T) | |
| Ethyl (2-Amino-4-thiazolyl)acetate |
| 94°C | |
| MFCD00005330 | |
| ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate | |
| CCOC(=O)CC1=CSC(N)=N1 | |
| 186.23 | |
| 186.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No