missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-phenylthiophene-3-carboxylate, 96%

Catalog No. AAH3186703
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3186703 1 g
AAH3186709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3186703 Supplier Thermo Scientific Chemicals Supplier No. H3186703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4815-36-5 | C13H13NO2S | 247.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4815-36-5
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
MDL Number MFCD00126391
InChI Key WYTHTMKMOSPACP-UHFFFAOYSA-N
Synonym 2-amino-4-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-phenylthiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4-phenyl-3-thiophenecarboxylate, thiophene-3-carboxylic acid, 2-amino-4-phenyl-, ethyl ester, amino-2 ethoxycarbonyl-3 phenyl-4 thiophene french, 3-thiophenecarboxylic acid, 2-amino-4-phenyl-, ethyl ester, ethyl 2-amino-4-phenyl-thiophene-3-carboxylate, 2-amino-4-phenyl-3-thiophenecarboxylic acid ethyl ester, cbmicro_024541, enamine_004212
PubChem CID 73242
IUPAC Name ethyl 2-amino-4-phenylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=CC=C2)N

Specifications

Melting Point 90°C to 94°C
Quantity 1 g
Formula Weight 247.32
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4-phenylthiophene-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only