missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-4-methylthiazole-5-carboxylate 98.0+%, TCI America™

Catalog No. p-7129210
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08981G 1 g
E08985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08981G Supplier TCI America Supplier No. E08981G
Only null left

Chemical Identifiers

CAS 7210-76-6
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00123414
InChI Key WZHUPCREDVWLKC-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-methylthiazole-5-carboxylate, ethyl2-amino-4-methylthiazole-5-carboxylate, 2-amino-4-methyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl 2-amino-4-methyl thiazole-5-carboxylate, ethyl 2-amino-4-methyl-thiazole-5-carboxylate, 2-amino-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-amino-4-methyl-5-ethoxycarbonyl thiazole, 2-amino-4-methyl-5-ethoxycarbonyl-thiazole, ethyl 2-amino-4-methyl-5-thiazolecarboxylate
PubChem CID 343747
IUPAC Name ethyl 2-amino-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=C(N)S1

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 1 g
Formula Weight 186.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-4-methylthiazole-5-carboxylate